Cobalt(II) oxalate dihydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Cobalt(II) oxalate dihydrate
CAS Number:5965-38-8
:
Formula:C₂CoO₄·2H₂O
MDL Number:MFCD03414148
:182.98 g/mol (146.95 anhy)
Merck Index:14,2445
Melting Pt:229…231 °C (dec.)
Density:3.021
Storage Temperature:Ambient
EINECS:212-409-3
UN:3288
ADR:6.1,III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together