Membrane filters, PORAFIL® PC
Supplier: MACHEREY-NAGEL
Certificates
About this item
Polycarbonate membranes are mainly used for the determination of AOX.
Frequently Bought Together
VWR® Weighing Paper, Grade 531
Supplier: VWR Collection
Syringe filters, Acrodisc®, Supor® membrane
Supplier: Cytiva (Formerly Pall Lab)
VWR® Extraction Thimbles, Grade 501
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Syringe Filters
Supplier: VWR Collection
VWR®, Bottle-Top Vacuum Filtration Systems, PES
Supplier: VWR Collection
VWR®, Glass Fibre Filters
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Pressure sprayer, LaboPlast®
Supplier: BURKLE
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Qualitative Filter Papers
Supplier: VWR Collection
Vacuum filtration units, Rapid-Flow™ filters MF 75, Nalgene®
Supplier: Thermo Fisher Scientific
Glass microfibre filters without binder, grade GF/C, Whatman™
Supplier: Whatman products (Cytiva)
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection