Membrane filters, PORAFIL® PE
Polyester membranes are particularly suited for fine filtration, dust analysis, aerosol analysis and ultra-purification of solvents.
- Hydrophilic
Frequently Bought Together
Polyester membranes are particularly suited for fine filtration, dust analysis, aerosol analysis and ultra-purification of solvents.