Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12092-47-6
:
Formula:C₁₆H₂₄Cl₂Rh₂
MDL Number:MFCD00012415
:493.08 g/mol
:M-Clarity™ quality level = MQ100
EINECS:235-157-6
Frequently Bought Together