Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate, Sigma-Aldrich®
Catalog # SIAL334987-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
EINECS:460-220-1
CAS Number:207124-65-0
Formula:C₁₆H₂₄BF₄Rh·H₂O
MDL Number:MFCD00167214