Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Piperonyl butoxide
Piperonyl butoxide, Sigma-Aldrich®
Piperonyl butoxide, Sigma-Aldrich®
SIAL291102-100ML
:
Merck
:
Piperonyl butoxide, Sigma-Aldrich®
SIAL291102-100ML
:
Merck
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
338.44 g/mol
:
9,III
:
M-Clarity™ quality level = MQ200
:
200-076-7
:
3082
:
51-03-6
:
C₁₉H₃₀O₅
:
MFCD00005842