
Tri-tert-butylphosphonium tetrafluoroborate ≥97%
Catalog # L19752.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:290.13 g/mol
Melting Pt:266…267 °C
:GHS07
Storage Temperature:Ambient
CAS Number:131274-22-1
:MFCD07189501
Formula:C₁₂H₂₈BF₄P
MDL Number:MFCD04039975