Order Entry
Slovenia
ContactUsLinkComponent
Benzyl chloride-[D₇] (99% D), Sigma-Aldrich®
Benzyl chloride-[D₇] (99% D), Sigma-Aldrich®
Catalog # SIAL217336-1G
Supplier:  Merck
CAS Number:  
Benzyl chloride-[D₇] (99% D), Sigma-Aldrich®
Catalog # SIAL217336-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Benzyl chloride-[²H₇]
 :
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
EINECS:261-790-2
UN:1738
 :GHS06
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:59502-05-5
Formula:(C₆D₅)CD₂Cl