Triethyl-2-phosphonopropionate
Catalog # MOLEM62899313
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:238.22 g/mol
EINECS:223-033-4
CAS Number:3699-66-9
Formula:C₉H₁₉O₅P
MDL Number:MFCD00009159