1-Bromo-2,5-bis(trifluoromethyl)benzene, Sigma-Aldrich®
Catalog # SIAL438782-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.01 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:7617-93-8
Formula:C₈H₃BrF₆
MDL Number:MFCD03094336