Order Entry
Slovenia
ContactUsLinkComponent
O-Methylhydroxylamine hydrochloride 98+%
Irritant
Corrosive
O-Methylhydroxylamine hydrochloride 98+%
Certificates
undefined
Irritant
Corrosive
O-Methylhydroxylamine hydrochloride 98+%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    148 to 153 °C
  • Titration with HClO4:
    98.0 to 102.0 %

Specification Test Results

Related Information

Frequently Bought Together