2-Phenylethanethiol 99%
:4410-99-5
:
:C₈H₁₀S
:MFCD00004891
:138.2300 g/mol
:97…100 °C (12 mmHg)
:1.03 g/cm³
:90 °C
:Ambient
:224-563-9
:3334
:9,III
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5590 to 1.5610 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together