Sodium bis(2-methoxyethoxy)aluminium hydride solution ≥60% (w/w) in toluene, Red-Al®, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium dihydrobis(2-methoxyethoxy)aluminate, SBAH, Sodium bis(2-methoxyethoxy)aluminum dihydride
CAS Number:22722-98-1
:
Formula:C₆H₁₆AlNaO₄
MDL Number:MFCD00011631
:202.16 g/mol
Density:1.036 g/mL (25 °C)
EINECS:245-178-2
UN:3399
ADR:4.3,I
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
:GHS07
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together