Tristearin (glycerol tristearate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Propane-1,2,3-triyl tristearate, Tristearin
CAS Number:555-43-1
:
Formula:C₅₇H₁₁₀O₆
MDL Number:MFCD00036230
Molecular Weight:891.4797 g/mol
EINECS:209-097-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together