
2-Chlorocyclohexanone ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:822-87-7
:
Formula:C₆H₉ClO
MDL Number:MFCD00001626
Boiling Pt:82…83 °C (10 mmHg)
Melting Pt:22…24 °C
Density:1.161
Flash Pt:82 °C (180 °F)
EINECS:212-505-5
UN:3335
ADR:9,
:Stabilised with calcium carbonate/magnesium oxide
:132.59 g/mol
:GHS07
Storage Temperature:Freezer
Related Information
Frequently Bought Together