(±)-α-Bromophenylacetic acid
Supplier: Molekula
Certificates
About this item
Synonyms: α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
CAS Number:4870-65-9
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00004206
Molecular Weight:215.0440 g/mol
EINECS:225-477-4
UN:3261
ADR:8 II
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together