EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16052-06-5
:
Formula:C₉H₂₀N₂O₄S
MDL Number:MFCD00006160
:M-Clarity™ quality level = MQ400
:252.33 g/mol
EINECS:240-198-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together