Vancomycin hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1404-93-9
:123409-00-7
Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
MDL Number:MFCD03613611
:M-Clarity™ quality level = MQ200
:1485.71 g/mol
Frequently Bought Together