Order Entry
Slovenia
ContactUsLinkComponent
Leupeptin, hemisulphate
Leupeptin, hemisulphate
Catalog # 786-051
Supplier:  G-Biosciences
CAS Number:  
Leupeptin, hemisulphate
Catalog # 786-051
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    25 mg
  • CAS No.:
    24365-47-7
  • Protein synonyms:
    Ac-Leu-Leu-Arg-al
  • Protein/peptide name:
    Leupeptin, hemisulfate
Specifications
About this item
Leupeptin, hemisulphate inhibits serine, plasmin, porcine kallikrein and cysteine proteases, including papain and cathepsin B. Does not inhibit chymotrypsin and thrombin