Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:34369-07-8
:
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MDL Number:MFCD00150755
:551.14 g/mol (anhy)
EINECS:213-579-1
Frequently Bought Together