Naphthol AS-BI phosphate disodium salt, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium 6-bromo-3-((2-methoxyphenyl)carbamoyl)naphthalen-2-yl phosphate
CAS Number:530-79-0
:
Formula:C₁₈H₁₃BrNNa₂O₆P
MDL Number:MFCD00150774
Molecular Weight:496.156 g/mol
:M-Clarity™ quality level = MQ200
EINECS:208-495-7
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together