Order Entry
Slovenia
ContactUsLinkComponent
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)
Catalog # ROTHCN22.2
Supplier:  Roth Carl
CAS Number:  
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)
Catalog # ROTHCN22.2
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :301.25 g/mol
Storage Temperature:Freezer
EINECS:206-716-1
CAS Number:369-07-3
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00063255