Order Entry
Slovenia
ContactUsLinkComponent
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) 99%
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) 99%
Certificates
undefined
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) 99%
Specification Test Results
  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 %
  • Specific optical rotation:
    -67° to -71° (20°C, 589 nm) (c=1, H2O)