TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥99% for biotechnology
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: 3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride
CAS Number:51805-45-9
:
Formula:C₉H₁₆ClO₆P
MDL Number:MFCD00145469
Molecular Weight:286.65 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity (TLC):>99%
- Solubility (50 mg/ml, water):pass
Specification Test Results
Frequently Bought Together