TRITON™ X-102 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
Catalog # SIALX102-500ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3082
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:9002-93-1