Pig Trypsin (from Pancreas)
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Pig
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin
- Enzyme activity:1.500 U/mg
Specifications
Frequently Bought Together