Order Entry
Slovenia
ContactUsLinkComponent
Apron, chrome-tanned, split leather
Apron, chrome-tanned, split leather
  113-08328
undefined
Apron, chrome-tanned, split leather
  113-08328

 

  • Closure type:
    Back fastening
  • Garment application:
    Industrial
  • Gender:
    Unisex
  • Colour:
    Light grey
  • L×W:
    1000×800 mm
  • Fabric:
    Cow split leather
  • Pk:
    1

 

 

Split leather apron made from top quality cow split leather with strong core leather straps.

L×W: approx. 100×80 cm

Colour: Light grey