Order Entry
Slovenia
ContactUsLinkComponent
Lotion soaps, Softaskin®
Lotion soaps, Softaskin®
Catalog # BRAU18168
Supplier:  B. Braun
undefined
Lotion soaps, Softaskin®
Catalog # BRAU18168
Supplier:  B. Braun

Specifications

  • Description:
    Lotion soap, Softaskin®, canister, 25 l
  • Seq:
    05
  • Pk:
    25 L

Specifications

About this item

Softaskin® is a mild wash lotion for sensitive skin.

  • Skin-friendly even with frequent wash
  • Skin neutral pH
  • Contains moisturiser allantoin to sooth irritated skin
  • Free of alkali, soap and colourants
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904