Chicken lysozyme (Muramidase) (from egg white)
Catalog # SIALL7773-50MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- CAS No.:12650-88-3
- Source:Egg white
- Species:Chicken
- Storage conditions:Freezer
- Enzyme name:Lysozyme (Muramidase)