Order Entry
Slovenia
ContactUsLinkComponent
Chicken lysozyme (Muramidase) (from egg white)
Chicken lysozyme (Muramidase) (from egg white)
Supplier:  Merck
Certificates
Chicken lysozyme (Muramidase) (from egg white)
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    12650-88-3
  • Source:
    Egg white
  • Species:
    Chicken
  • Storage conditions:
    Freezer
  • Enzyme name:
    Lysozyme (Muramidase)

Specifications

Related Information

Frequently Bought Together