0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Enzymes
Lysozyme (Muramidase)
Lysozyme (Muramidase)
PIER89833
:
Thermo Fisher Scientific
:
Lysozyme (Muramidase)
PIER89833
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
CAS No.:
9001-63-2
Enzyme name:
Lysozyme (Muramidase)