Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # ROCH11666681001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:4 L
- Description:SSC buffer solution 20X concentrate (saline-sodium citrate)
- M-Clarity quality level:MQ100