Buffer, PBS solution 10X concentrate (phosphate buffered saline)
PBS buffer solution 10X concentrate (phosphate buffered saline), Plastic bottle, 4 L
Supplier: ROCHE
Buffer, PBS solution 10X concentrate (phosphate buffered saline)
Catalog # ROCH11666789001
Supplier: ROCHE
CAS Number:
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:4 L
- Description:PBS buffer solution 10X concentrate (phosphate buffered saline)
- Cat. No.:ROCH11666789001
- M-Clarity quality level:MQ100
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



