Methyl-3-aminothiophene-2-carboxylate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:22288-78-4
:
Formula:C₆H₇NO₂S
MDL Number:MFCD00009765
:157.19 g/mol
:M-Clarity™ quality level = MQ200
EINECS:244-895-8
Frequently Bought Together