Dipropyleneglycol monomethyl ether (mixture of isomers), Sigma-Aldrich®
Catalog # SIAL484253-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:148.2001 g/mol
EINECS:252-104-2
CAS Number:34590-94-8
Formula:C₇H₁₆O₃
MDL Number:MFCD00059604