
Tetradecylphosphonic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4671-75-4
:
Formula:C₁₄H₃₁O₃P
MDL Number:MFCD00015834
:278.37 g/mol
EINECS:225-118-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together