0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Standards
Organic Standards
Pesticide reference standard, β-BHC 100 µg/ml in toluene, Dr Ehrenstorfer
Pesticide reference standard, β-BHC 100 µg/ml in toluene, Dr Ehrenstorfer
EHERXA14072000TO
:
EHRENSTORFER
:
Pesticide reference standard, β-BHC 100 µg/ml in toluene, Dr Ehrenstorfer
EHERXA14072000TO
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
1 mL
CAS No.:
319-85-7
Description:
β-BHC 100 µg/ml in toluene
Formula:
C₆H₆Cl₆
Description:
β-BHC