Safety boots, Wellingtons, Nora
Supplier: UVEX
Certificates
About this item
High PVC safety boots are anti-static, resistant to oil, grease, fuel, acids, alkalis and disinfectants.
- Anatomical shape prevents foot fatigue
- Long shaft can be shortend to meet individual requirements
- Toe and heel reinforcement
- Ankle protection and ribbed heel with kick-off tab to assist removal
- Protective steel toecap (200 J)
- Puncture resistant steel midsole
Colour: yellow or white.
Shaft height: 330 mm.
Certifications: EN 20345:2011; S5, SRA