


Tris(2-aminoethyl)amine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4097-89-6
:
Formula:C₆H₁₈N₄
MDL Number:MFCD00008177
:146.24 g/mol
Boiling Pt:265 °C (760 mmHg)
Melting Pt:–16 °C
Density:0.977 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:223-857-4
UN:2922
ADR:8,II
:GHS06
:GHS05
:GHS08
Specification Test Results
- Appearance:Clear colorless to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4960 to 1.4980 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together