Pesticide reference standard, Cadusafos 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL10934000CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:95465-99-9
- Description:Cadusafos 10 µg/ml in cyclohexane
- Formula:C₁₀H₂₃O₂PS₂
- Description:Cadusafos