Order Entry
Slovenia
ContactUsLinkComponent
 
7-Hydroxy-2-tetralone
Catalog # MOLEM73492513
Supplier:  Molekula
CAS Number:  
7-Hydroxy-2-tetralone
Catalog # MOLEM73492513
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 7-Hydroxy-3,4-dihydro-1H-naphthalen-2-one
 :
 :162.19 g/mol
CAS Number:37827-68-2
Formula:C₁₀H₁₀O₂
MDL Number:MFCD04038657