Order Entry
Slovenia
ContactUsLinkComponent
 
Gel filtration media, Sephadex™ G-50
 :  Cytiva
 
undefined
Gel filtration media, Sephadex™ G-50
 :  Cytiva
 

 

  • pH range:
    2 - 10
  • Matrix:
    Cross-linked dextran
  • Fractionation range:
    1 × 102 - 5 ×103 dextrans, 1 × 103 - 3 × 104 globular proteins
  • Storage temperature:
    4 - 30 °C
  • Description:
    Sephadex™ G-50 Superfine

 

Frequently Bought Together