Order Entry
Slovenia
ContactUsLinkComponent
Absorption granules for chemical spillages, Uni-Safe
Absorption granules for chemical spillages, Uni-Safe
Catalog # BAST271500
undefined
Absorption granules for chemical spillages, Uni-Safe
Catalog # BAST271500
Specifications
  • Description:
    Chemical spillage absorption granules, UNI-SAFE, in 5 kg bucket
  • Seq:
    02
  • Pk:
    5 kg
Specifications
About this item
Spillage granules for rapid absorbance of liquid chemicals, acids, alkalis, oil, gasoline and similar substances.
  • Simply add the granules over the spillage for safe treatment of liquids, it binds up to 75 times its own weight
  • This results in only small quantities of waste disposal and therefore reduced recycling costs
  • In addition, with UNI-SAFE there are no negative impacts on the environment