Order Entry
Slovenia
Orders LinkContactUsLinkComponent
di-Lithium oxalate ≥99%

di-Lithium oxalate ≥99%

di-Lithium oxalate ≥99%
Certificates
di-Lithium oxalate ≥99%
di-Lithium oxalate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 553-91-3 Molecular Formula: Li2C2O4 Formula Weight: 101.96 Physical Form: Powder Density: 2.12 Melting Point: dec. Solubility: Soluble in water Merck Reference: 14,5537 UN No.: 3288 MDL No.: MFCD00040596

Synonyms:  Dilithium oxalate, Lithium oxalate, Lithiumoxalate
  • CAS Number:553-91-3
  • Formula:C₂Li₂O₄
  • MDL Number:MFCD00040596
  • Molecular Weight:101.9 g/mol
  • Merck Index:14,5537
  • Melting Pt:Dec.
  • Density:2.12
  • Storage Temperature:Ambient
  • EINECS:209-054-1
  • UN:3288
  • ADR:6.1,III
Product Family Options

Product Information