Boron standard solution, 10000 mg/L B in ammonium hydroxide solution (max. 1%) standard for AAS
Catalog # PROOU-IAA-005
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:<1% NH₄OH
- Pack type:Glass bottle
- Pk:10 mL
- Environmentally Preferable:
- Element:Boron
- Symbol:B