Tubing connectors, T-shaped, BOLA
Supplier: Bohlender
Certificates
About this item
Tubing connectors, T-shaped, made of PTFE, for flexible tubings.
- Universal chemical resistance
- Temperature resistance: from −200 to +250 °C
- Sutable for vacuum applications
- FDA conform
Frequently Bought Together