Pesticide reference standard, 2-Imidazolidinethione 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL13330000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:96-45-7
- Description:2-Imidazolidinethione 10 µg/ml in acetonitrile
- Formula:C₃H₆N₂S
- Description:2-Imidazolidinethione