

Silver(I) oxide ≥99%, TECHNICAL
21584.134
:
- Pack type:Plastic bottle for solids
- Pk:25 g
- Seq:3308
- Assay:Min. 99 %
:
:231.74 g/mol
:12,08665
:800…830 °C
:7.14 g/cm³ (20 °C)
:Refrigerator
:243-957-1
:1479
:5.1,II
:GHS03
:GHS05
:20667-12-3
:Ag₂O
:MFCD00003404