


Antimony(III) chloride 99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-91-9
:
Formula:SbCl₃
MDL Number:MFCD00011212
:228.12 g/mol
Merck Index:13,00713
Boiling Pt:223.5 °C (1013 hPa)
Melting Pt:73 °C
Density:3,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-047-2
UN:1733
ADR:8,II
:GHS05
:GHS09
:GHS07
Specification Test Results
- Appearance:Colorless to white to off-white Adhering crystals and/or chunks (May have a pink hue)
- Titration Iodimetric:≥99.4 %
Specification Test Results
Frequently Bought Together