Disodium oxide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-59-3
:
Formula:Na₂O
MDL Number:MFCD00046201
:61.98 g/mol
Merck Index:14,8651
Melting Pt:>400 °C (dec.)
Density:2.27
Storage Temperature:Ambient
EINECS:215-208-9
UN:3085
ADR:5.1,I
:GHS03
:GHS05
Related Information
Frequently Bought Together